Order Entry
Northern Ireland
ContactUsLinkComponent
Tributyl citrate for synthesis, Sigma-Aldrich®
Tributyl citrate for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Tributyl citrate for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    (20 °C) insoluble
  • Melting Point:
    -20 °C
  • Molar Mass:
    360.44 g/mol
  • Boiling Point:
    169 - 171 °C (1 hPa)
  • Flash Point:
    184 °C
  • Refractive Index:
    1.443 (25 °C, 589 nm)
  • Density:
    1.04 g/cm3 (20 °C)

Specification Test Results

Related Information

Frequently Bought Together