

Diethyl oxalate for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:95-92-1
:
Formula:C₆H₁₀O₄
MDL Number:MFCD00009119
:146.14 g/mol
Merck Index:13,03152
Boiling Pt:185,7 °C (1013 hPa)
Melting Pt:–41 °C
Density:1,0785 g/cm³ (20 °C)
EINECS:202-464-1
UN:2525
ADR:6.1,III
:M-Clarity™ quality level = MQ200
Flash Pt:76 °C
:GHS05
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:(20 °C) (slow decomposition)
- Melting Point:-41 °C
- Molar Mass:146.14 g/mol
- Boiling Point:185.7 °C (1013 hPa)
- Vapor Pressure:0.27 hPa (20 °C)
- Flash Point:75 °C
- Refractive Index:1.4096 (20 °C, 589 nm)
- Explosion Limit:0.42 - 2.67 %(V)
- Density:1.08 g/cm3 (20 °C)
- Ignition Point:410 °C
Specification Test Results
Related Information
Frequently Bought Together