
Diethyl oxalate ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:95-92-1
:
Formula:C₆H₁₀O₄
MDL Number:MFCD00009119
:146.14 g/mol
Merck Index:14,3125
Boiling Pt:184…186 °C
Melting Pt:–41 °C
Density:1,077
Flash Pt:75 °C (167 °F)
Storage Temperature:Ambient
EINECS:202-464-1
UN:2525
ADR:6.1,III
:GHS07
Related Information
Frequently Bought Together