
Dimethyl succinate for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:106-65-0
:
Formula:C₆H₁₀O₄
MDL Number:MFCD00008466
:146.14 g/mol
Merck Index:12,09038
Boiling Pt:200 °C (1013 hPa)
Melting Pt:18…19 °C
Density:1,117 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
EINECS:203-419-9
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:75 g/l (20 °C)
- Melting Point:18 - 19 °C
- Molar Mass:146.14 g/mol
- Boiling Point:196 °C (1013 hPa)
- Vapor Pressure:0.2 hPa (20 °C)
- Flash Point:94 °C
- Refractive Index:1.419 (20 °C, 589 nm)
- Explosion Limit:1.0 - 8.5 %(V)
- Density:1.12 g/cm3 (20 °C)
- Ignition Point:365 °C
Specification Test Results
Frequently Bought Together