
Methyl octanoate for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:111-11-5
:
Formula:C₉H₁₈O₂
MDL Number:MFCD00009551
:158.24 g/mol
Boiling Pt:190 °C (1013 hPa)
Melting Pt:–36,7 °C
Density:0,877 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Flash Pt:68 °C
EINECS:203-835-0
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:0.064 g/l (20 °C)
- Melting Point:-40 °C
- Molar Mass:158.24 g/mol
- Boiling Point:193 °C (1013 hPa)
- Vapor Pressure:1.33 hPa (34.2 °C)
- Flash Point:68 °C
- Density:0.877 g/cm3 (20 °C)
Specification Test Results
Frequently Bought Together