

Ethyl formate for synthesis
Catalog # 8.00891.9045
Supplier: Merck
CAS Number:
Specifications
- Pk:45 kg
- Seq:4388
Specifications
About this item
:
:74.08 g/mol
Merck Index:13,03841
Boiling Pt:148 °C (1013 hPa)
Melting Pt:–2 °C
Density:0,9208 g/cm³ (20 °C)
Flash Pt:–20 °C
Storage Temperature:Ambient
EINECS:203-721-0
UN:1190
ADR:3,II
:GHS02
:GHS07
CAS Number:109-94-4
Formula:C₃H₆O₂
MDL Number:MFCD00003294