
Acetamide for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:60-35-5
:
Formula:C₂H₅NO
MDL Number:MFCD00008023
:59.07 g/mol
Merck Index:13,00044
Boiling Pt:221,1 °C (1013 hPa)
Melting Pt:78…81 °C
Density:1,14 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
EINECS:200-473-5
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Solubility:2200 g/l (20 °C)
- Melting Point:78 - 81 °C
- Molar Mass:59.06 g/mol
- Boiling Point:221 - 222 °C (1013 hPa)
- Vapor Pressure:1.33 hPa (65 °C)
- Density:1.159 g/cm3 (20 °C)
Specification Test Results
Frequently Bought Together