


N,N-Dimethylformamide, Supelco®
72438-5ML-F
: Merck
:
- Pack type:Glass bottle
- Pk:5 mL
- Seq:2210
:
:73.09 g/mol
:13,03269
:153 °C (1013 hPa)
:–61 °C
:0,949 g/cm³ (20 °C)
:57,5 °C (closed cup)
:Ambient
:200-679-5
:2265
:3,III
:M-Clarity™ quality level = MQ100
:01-2119475605-32
:GHS02
:GHS08
:GHS07
:68-12-2
:HCON(CH₃)₂
:MFCD00003284