

N,N-Dimethylacetamide ≥99.5%, AnalaR NORMAPUR® analytical reagent
Catalog # 103646E
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:2,5 L
- Seq:5096
Specifications
Specification Test Results
- Assay:Min. 99.5 %
- Insolubility in water:Passes test
- IR Spectrum:Passes test
- Acidity:Max. 0.02 meq/g
- Colouration:Max. 10 APHA
- Non-volatile residue:Max. 100 ppm
- Water:Max. 0.1 %
- Cl (Chloride):Max. 10 ppm
- SO4 (Sulphate):Max. 10 ppm
- Cu (Copper):Max. 1 ppm
- Fe (Iron):Max. 1 ppm
- Pb (Lead):Max. 1 ppm
Specification Test Results
About this item
:
:87.12 g/mol
Merck Index:13,03254
Boiling Pt:166,1 °C (1013 hPa)
Melting Pt:–20 °C
Density:0,937 g/cm³ (20 °C)
Flash Pt:70 °C
Storage Temperature:Ambient
EINECS:204-826-4
UN:2810
ADR:6.1,III
REACH:01-2119459339-27
:GHS08
:GHS07
CAS Number:127-19-5
Formula:CH₃CON(CH₃)₂
MDL Number:MFCD00008686