3-Oxoglutaric acid for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:542-05-2
:
Formula:C₅H₆O₅
MDL Number:MFCD00002711
:146.1 g/mol
Merck Index:13,00069
Boiling Pt:323 °C (1013 hPa)
Melting Pt:138 °C
Density:1,55 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Freezer
EINECS:208-797-9
Specification Test Results
- Solubility:(20 °C) soluble
- Molar Mass:146.10 g/mol
- Bulk Density:400 kg/m3
- Synonyms:ß-Ketoglutaric acid, Acetone-1,3-dicarboxylic acid
- Hill Formula:C5H6O5
- Chemical Formula:HOOCCH2COCH2COOH
Specification Test Results
Frequently Bought Together