
Ethyl acetoacetate for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:141-97-9
:
Formula:C₆H₁₀O₃
MDL Number:MFCD00009199
Merck Index:13,03793
Boiling Pt:180,8 °C (1013 hPa)
Melting Pt:–45 °C
Density:1,0188 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Flash Pt:65 °C
:130.14 g/mol
EINECS:205-516-1
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:116 g/l (20 °C)
- Melting Point:-40 °C
- Molar Mass:130.14 g/mol
- Boiling Point:180.0 °C (1013 hPa)
- Vapor Pressure:1 hPa (20 °C)
- Flash Point:65 °C
- Refractive Index:1.419 (20 °C, 589 nm)
- Explosion Limit:1.0 - 54 %(V)
- Density:1.03 g/cm3 (20 °C)
- pH:4.0 (110 g/l, H2O, 20 °C)
- Ignition Point:280 °C
Specification Test Results
Frequently Bought Together