Ethyl pyruvate for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:617-35-6
:
Formula:C₅H₈O₃
MDL Number:MFCD00009123
:116.12 g/mol
Boiling Pt:144…147 °C (1013 hPa)
Melting Pt:–50 °C
Density:1,05 g/cm³ (20 °C)
EINECS:210-511-2
ADR:3,III
:M-Clarity™ quality level = MQ200
UN:3272
:GHS02
Storage Temperature:Refrigerator
Specification Test Results
- Solubility:10 g/l (20 °C)
- Melting Point:-58 °C
- Molar Mass:116.11 g/mol
- Boiling Point:144 - 146 °C (1013 hPa)
- Flash Point:52 °C
- Refractive Index:1.4056 (20 °C, 589 nm)
- Density:1.05 g/cm3 (20 °C)
- pH:2.2 (300 g/l, H2O, 20 °C)
- Hazard Class:3
- Hazard Class Description:Flammable Liquid
Specification Test Results
Frequently Bought Together