Ethyl pyruvate 98%
Supplier: Thermo Fisher Scientific
About this item
Synonyms: Ethyl 2-oxopropanoate, 2-Oxopropionic acid ethyl ester, α-Ketopropionic acid ethyl ester, Pyruvic acid ethyl ester, 2-Ketopropionic acid ethyl ester
- CAS Number:617-35-6
- Formula:C₅H₈O₃
- MDL Number:MFCD00009123
- Molecular Weight:116.12 g/mol
- Boiling Pt:144…147 °C (1013 hPa)
- Melting Pt:–58 °C
- Density:1,0453 g/cm³
- Flash Pt:45 °C
- Storage Temperature:Refrigerator
- EINECS:210-511-2
- UN:3272
- ADR:3,III
2 Options Available Below
Product Details & Documents
Synonyms: Ethyl 2-oxopropanoate, 2-Oxopropionic acid ethyl ester, α-Ketopropionic acid ethyl ester, Pyruvic acid ethyl ester, 2-Ketopropionic acid ethyl ester
- CAS Number:617-35-6
- Formula:C₅H₈O₃
- MDL Number:MFCD00009123
- Molecular Weight:116.12 g/mol
- Boiling Pt:144…147 °C (1013 hPa)
- Melting Pt:–58 °C
- Density:1,0453 g/cm³
- Flash Pt:45 °C
- Storage Temperature:Refrigerator
- EINECS:210-511-2
- UN:3272
- ADR:3,III
Documents
ACRO11894
SDS
Specifications for Product Family
Specification Test Results
- Appearance:Clear light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4040 to 1.4060 (20°C, 589 nm)
- GC:≥97.5 %
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



