Order Entry
Northern Ireland
ContactUsLinkComponent
3-Hydroxypropiononitrile for synthesis, Sigma-Aldrich®
3-Hydroxypropiononitrile for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
3-Hydroxypropiononitrile for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    1000 g/l (20 °C)
  • Melting Point:
    -46 °C
  • Molar Mass:
    71.08 g/mol
  • Boiling Point:
    228 °C (1013 hPa)
  • Vapor Pressure:
     - 0.01 hPa (20 °C)
  • Flash Point:
    128 °C DIN 51758
  • Refractive Index:
    1.4241 (25 °C, 589 nm)
  • Density:
    1.05 g/cm3 (20 °C)
  • pH:
    3.0 - 4.5 (H2O, 20 °C) (undiluted)

Specification Test Results

Related Information

Frequently Bought Together