

Trichloroacetonitrile for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:545-06-2
:
Formula:C₂Cl₃N
MDL Number:MFCD00001842
:144.39 g/mol
Merck Index:12,09757
Boiling Pt:85,7 °C (1013 hPa)
Melting Pt:–44 °C
Density:1,44 g/cm³ (20 °C)
UN:3276
ADR:6.1,III
:M-Clarity™ quality level = MQ200
EINECS:208-885-7
:GHS06
:GHS09
Storage Temperature:Ambient
Frequently Bought Together