


Propylene oxide for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:75-56-9
:
Formula:C₃H₆O
MDL Number:MFCD00005126
:58.08 g/mol
Merck Index:12,08041
Boiling Pt:34,3 °C (1013 hPa)
Melting Pt:–112 °C
Density:0,83 g/cm³ (20 °C)
EINECS:200-879-2
UN:1280
ADR:3,I
:M-Clarity™ quality level = MQ200
Flash Pt:–37 °C
:GHS02
:GHS06
:GHS08
Storage Temperature:Refrigerator
Specification Test Results
- Solubility:405 g/l (20 °C)
- Melting Point:-112 °C
- Molar Mass:58.07 g/mol
- Boiling Point:34.3 °C (1013 hPa)
- Vapor Pressure:585 hPa (20 °C)
- Flash Point:-37 °C
- Refractive Index:1.366 (20 °C, 589 nm)
- Explosion Limit:1.9 - 36 %(V)
- Density:0.83 g/cm3 (20 °C)
- Ignition Point:430 °C
- Synonyms:1,2-Epoxypropane, Methyloxirane
- Hill Formula:C3H6O
Specification Test Results
Frequently Bought Together