

1,2-Epoxyoctan ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2984-50-1
:
Formula:C₈H₁₆O
MDL Number:MFCD00005157
:128.21 g/mol
Boiling Pt:167 °C
Density:0,838
Flash Pt:37 °C (98 °F)
Storage Temperature:Refrigerator
EINECS:221-047-5
UN:1993
ADR:3,III
:GHS02
:GHS07
Related Information
Frequently Bought Together