1,2-Epoxyhexane 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1436-34-6
:
Formula:C₆H₁₂O
MDL Number:MFCD00005154
:100.16 g/mol
Boiling Pt:118…120 °C (760 mmHg)
Density:0,83 g/cm³
Flash Pt:15 °C
Storage Temperature:Refrigerator
EINECS:215-864-6
UN:1993
ADR:3,II
:GHS07
:GHS02
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4040 to 1.4070 (20°C, 589 nm)
- GC:≥96.0 %
Specification Test Results
Frequently Bought Together