
Dibromomethane
Catalog # 47799.
Supplier: Merck
CAS Number:
Specifications
- Pk:1
- Seq:0001
Specifications
About this item
:
:173.83 g/mol
Merck Index:12,06138
Boiling Pt:97 °C (1013 hPa)
Melting Pt:–53 °C
Density:2,497 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-824-2
UN:2664
ADR:6.1,III
:GHS07
CAS Number:74-95-3
Formula:Br₂CH₂
MDL Number:MFCD00000168