Order Entry
Northern Ireland
ContactUsLinkComponent
 
 
 
2,4-Dimethyl-1,3-pentadiene 98%
 
undefined
 
 
2,4-Dimethyl-1,3-pentadiene 98%

 

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4400 to 1.4420 (20°C, 589 nm)
  • GC:
    ≥97.5 %

 

 

Frequently Bought Together