(R)-(+)-1-Phenylethanol ≥99%, ee 97%, ChiPros®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1517-69-7
:
Formula:C₈H₁₀O
MDL Number:MFCD00064263
:122.17 g/mol
Boiling Pt:88…89 °C (10 mmHg)
Melting Pt:9…11 °C
Density:1,012
Flash Pt:85 °C (185 °F)
Storage Temperature:Ambient
UN:2937
ADR:6.1,III
:GHS05
:GHS07
Related Information
Frequently Bought Together