Order Entry
Northern Ireland
Orders LinkContactUsLinkComponent
L(+)-Histidine

L(+)-Histidine

Glass bottle for solids, 1 kg
Catalog # MOLE16752688-1KG
Marketsource
Supplier:   Molekula
L(+)-Histidine
Catalog # MOLE16752688-1KG
Supplier:  Molekula
Marketsource
CAS Number:  71-00-1
L(+)-Histidine
L(+)-Histidine

Glass bottle for solids, 1 kg

CAS Number:  71-00-1
Price
£555.00
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  L-3-Imidazol-4-ylalanine, (S)-(+)-Histidine, (S)-2-Amino-3-(4-imidazolyl)propionic acid, H-His-OH, NSC 137773, L-Histidine
  • CAS Number:71-00-1
  • Formula:C₆H₉N₃O₂
  • MDL Number:MFCD00064315
  • Molecular Weight:155.16 g/mol
  • Merck Index:13,04741
  • Melting Pt:272…273 °C
  • Density:1,44 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-745-3
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.