L(+)-Histidine ≥98%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Synonyms: L-3-Imidazol-4-ylalanine, (S)-(+)-Histidine, (S)-2-Amino-3-(4-imidazolyl)propionic acid, H-His-OH, NSC 137773, L-Histidine
- CAS Number:71-00-1
- Formula:C₆H₉N₃O₂
- MDL Number:MFCD00064315
- Molecular Weight:155.16 g/mol
- Merck Index:14,4720
- Melting Pt:∼ 281 °C (dec.)
- Density:1,44 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:200-745-3
3 Options Available Below
Product Details & Documents
Synonyms: L-3-Imidazol-4-ylalanine, (S)-(+)-Histidine, (S)-2-Amino-3-(4-imidazolyl)propionic acid, H-His-OH, NSC 137773, L-Histidine
- CAS Number:71-00-1
- Formula:C₆H₉N₃O₂
- MDL Number:MFCD00064315
- Molecular Weight:155.16 g/mol
- Merck Index:14,4720
- Melting Pt:∼ 281 °C (dec.)
- Density:1,44 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:200-745-3
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



