Parteck® SI 400 LEX (Sorbitol), EMPROVE® EXPERT Ph. Eur., BP, NF, JP suitable for use as excipient, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Certificates
About this item
CAS Number:50-70-4
:98201-93-5
Formula:HOH₂C(CH(OH))₄CH₂OH
MDL Number:MFCD00004708
Merck Index:13,08797
Boiling Pt:295 °C (1013 hPa)
Melting Pt:94…98 °C
Density:1,524 g/cm³ (20 °C)
Flash Pt:>100 °C
:182.17 g/mol
Storage Temperature:Ambient
EINECS:200-061-5
Specification Test Results
- Assay (HPLC, calc.on anhydrous substance):97.0 - 100.5 %
- Identity:passes test
- pH-value (10 %; water):3.5 - 7.0
- Appearance of solution (25 %; water):passes test
- Spec. rotation (α20/D, 10 %, borate complex, calc. on anhydrous substance):+4.0 - +7.0 °
- Conductivity (25°C) (20 %; water):≤ 20
- Chloride (Cl):≤ 0.002 %
- Sulfate (SO₄):≤ 0.006 %
- Heavy metals (as Pb):≤ 5 ppm
- As (Arsenic):≤ 1.3 ppm
- Ni (Nickel) (#):≤ 1 ppm
- Pb (Lead):≤ 0.5 ppm
- Related substances (HPLC, EP) (Major impurity (Mannitol)):≤ 2.0 %
- Related substances (HPLC, EP) (Sum of all impurities):≤ 3.0 %
- Reducing sugars after hydrolysis/total sugar (as glucose):≤ 0.8
- Reducing sugars (as glucose):≤ 0.11 %
- Sulfated ash:≤ 0.02 %
- Loss on drying:≤ 2.0 %
- Water:≤ 1.5 %
- Residual solvents (ICH Q3C):excluded by manufacturing process
- Microbiological test:passes test
- Colony count (aerobic bacteria (TAMC)):≤ 10²
- Colony count (Yeasts and moulds (TYMC)):≤ 10²
- Salmonella spp.:in 10g Substance not detectable
- E.coli:in 1g substance not detectable
- Pseudomonas aeruginosa:in 1g substance not detectable
- Staphylococcus aureus:in 1g substance not detectable
- Candida albicans:In 1g substance not detectable
- Endotoxins:≤ 1
Specification Test Results
Related Information
Frequently Bought Together