(S)-(+)-1,2-Propanediol ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4254-15-3
:
Formula:C₃H₈O₂
MDL Number:MFCD00004539
:76.1 g/mol
Merck Index:14,7855
Boiling Pt:186…188 °C
Density:1,040
Flash Pt:107 °C (224 °F)
Storage Temperature:Ambient
Related Information
Frequently Bought Together