Pesticide reference standard, β-BHC 100 µg/ml in methanol
Catalog # ACCUP-011S
Supplier: AccuStandard
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:319-85-7
- Description:β-BHC 100 µg/ml in methanol
- Formula:C₆H₆Cl₆
- Description:β-BHC