Pesticide reference standard, (±)-α-BHC 100 µg/ml in methanol
Catalog # ACCUP-010S
Supplier: AccuStandard
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:319-84-6
- Description:(±)-α-BHC 100 µg/ml in methanol
- Formula:C₆H₆Cl₆
- Description:(±)-α-BHC