Order Entry
Northern Ireland
ContactUsLinkComponent
Pesticide reference standard, δ-BHC 100 µg/ml in methanol
Pesticide reference standard, δ-BHC 100 µg/ml in methanol
Catalog # ACCUP-012S
Supplier:  AccuStandard
CAS Number:  
Pesticide reference standard, δ-BHC 100 µg/ml in methanol
Catalog # ACCUP-012S
Supplier:  AccuStandard
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    319-86-8
  • Description:
    δ-BHC 100 µg/ml in methanol
  • Formula:
    C₆H₆Cl₆
  • Description:
    δ-BHC

Specifications

About this item