Organic reference standard, γ-Hexabromocyclododecane 50 µg/ml in toluene, CIL
PROOCILULM48361.2
:
- Pack type:Glass bottle
- Pk:1,2 mL
- CAS No.:134237-52-8
- Description:γ-Hexabromocyclododecane 50 µg/ml in toluene
- Formula:C₁₂H₁₈Br₆
- Description:γ-Hexabromocyclododecane