Order Entry
Northern Ireland
ContactUsLinkComponent
Organic reference standard, 2,3,3',4,4',5,5',6-Octachlorobiphenyl (PCB No. 205) 35 µg/ml in isooctane, CIL
Organic reference standard, 2,3,3',4,4',5,5',6-Octachlorobiphenyl (PCB No. 205) 35 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-205
CAS Number:  
Organic reference standard, 2,3,3',4,4',5,5',6-Octachlorobiphenyl (PCB No. 205) 35 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-205
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    74472-53-0
  • Description:
    2,3,3',4,4',5,5',6-Octachlorobiphenyl (PCB No. 205) 35 µg/ml in isooctane
  • Formula:
    C₁₂H₂Cl₈
  • Description:
    2,3,3',4,4',5,5',6-Octachlorobiphenyl (PCB No. 205)

Specifications

About this item