Order Entry
Northern Ireland
ContactUsLinkComponent
Organic reference standard, 3-Chlorobiphenyl (PCB No. 2) 35 µg/ml in isooctane, CIL
Organic reference standard, 3-Chlorobiphenyl (PCB No. 2) 35 µg/ml in isooctane, CIL
  PROOCIL-PCB-2
 :  
Organic reference standard, 3-Chlorobiphenyl (PCB No. 2) 35 µg/ml in isooctane, CIL
  PROOCIL-PCB-2
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    2051-61-8
  • Description:
    3-Chlorobiphenyl (PCB No. 2) 35 µg/ml in isooctane
  • Formula:
    C₁₂H₉Cl
  • Description:
    3-Chlorobiphenyl (PCB No. 2)