£0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Northern Ireland
ContactUsLinkComponent
D(+)-Malic acid
D-(+)-Malic acid ≥98%
D-(+)-Malic acid ≥98%
:
Thermo Fisher Scientific
D-(+)-Malic acid ≥98%
:
Thermo Fisher Scientific
:
636-61-3
:
:
C₄H₆O₅
:
MFCD00004245
:
134.09 g/mol
:
14,5707
:
322 °C (1013 hPa)
:
100…104 °C
:
1,60
:
Ambient
:
211-262-2
:
GHS05
:
GHS07
msds_ALFAA11688
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years
Frequently Bought Together
N,N-Dimethylformamide, EMPLURA®, Supelco®
: Merck
Petroleum spirit, 40…60 °C, GPR RECTAPUR®
: VWR Chemicals
Dichloromethane ≥99.8% stabilised, PESTINORM® for pesticide residue analysis
: VWR Chemicals
Petroleum spirit, 40…60 °C, AnalaR NORMAPUR® ACS analytical reagent (max. 0.01% aromatic hydrocarbons)
: VWR Chemicals
Acetone ≥99.7%, PESTINORM® for pesticide residue analysis
: VWR Chemicals
Tetrahydrofuran ≥99% stabilised, GPR RECTAPUR®
: VWR Chemicals
Butyl acetate ≥99.5% (by GC), EMSURE® for analysis, Supelco®
: Merck
n-Hexane ≥99.0%, SPECTRONORM® for spectroscopy
: VWR Chemicals
Ethyl acetate, extra dry over Molecular Sieve 99.9%, AcroSeal®
: Thermo Fisher Scientific
N-Methyl-2-pyrrolidone (NMP) 99.5% for HPLC
: Thermo Fisher Scientific
Succinic acid ACS
: Thermo Fisher Scientific
Diiodomethane 99+% stabilised
: Thermo Fisher Scientific
Water, extra pure, Deionized
: Thermo Fisher Scientific
Ethylene glycol ≥99%
: Thermo Fisher Scientific
Methanol bio-based ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Petroleum spirit, 30…40 °C, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Water suitable for ion chromatography, Supelco®
: Merck
N,O-Bis(trimethylsilyl)trifluoroacetamide with 1% trimethylchlorosilane 99% (excluding TMCS) (by GC), LiChropur™ for GC derivatisation, Supelco®
: Merck