L(+)-Aspartic acid for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:56-84-8
:
Formula:C₄H₇NO₄
MDL Number:MFCD00002616
:133.1 g/mol
Merck Index:13,00845
Boiling Pt:311 °C (1013 hPa)
Melting Pt:251 °C
Density:1,6613 g/cm³ (13 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:200-291-6
Specification Test Results
- Solubility:4 g/l (20 °C)
- Melting Point:269 - 271 °C (decomposition)
- Molar Mass:133.1 g/mol
- Bulk Density:430 kg/m3
- pH:2.5 - 3.5 (4 g/l, H2O, 20 °C)
Specification Test Results
Related Information
Certificates
Search by Lot Numbermsds_816003
SDS
ProductPage_816003 ProductPage_816003
Product specification
Frequently Bought Together