L(+)-Aspartic acid
Catalog # A1701.1000
Supplier: PanReac AppliChem
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
About this item
:
:133.1 g/mol
Merck Index:13,00845
Boiling Pt:311 °C (1013 hPa)
Melting Pt:251 °C
Density:1,6613 g/cm³ (13 °C)
Storage Temperature:Ambient
EINECS:200-291-6
CAS Number:56-84-8
Formula:C₄H₇NO₄
MDL Number:MFCD00002616