L(+)-Aspartic acid ≥98%, high purity
0192-500G
:
- Pack type:Plastic bottle for solids
- Pk:500 g
- Seq:3645
- Loss on Drying:< = 0.2 %
- Purity:>= 98 %
:
:133.1 g/mol
:13,00845
:311 °C (1013 hPa)
:251 °C
:1,6613 g/cm³ (13 °C)
:Ambient
:200-291-6
:56-84-8
:C₄H₇NO₄
:MFCD00002616