Order Entry
Northern Ireland
ContactUsLinkComponent
 
Organic reference standard, 2,3,3',4,5'-Pentachlorobiphenyl (PCB No. 108) 35 µg/ml in isooctane, CIL
  PROOCIL-PCB-108
 :  
Organic reference standard, 2,3,3',4,5'-Pentachlorobiphenyl (PCB No. 108) 35 µg/ml in isooctane, CIL
  PROOCIL-PCB-108
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    70362-41-3
  • Description:
    2,3,3',4,5'-Pentachlorobiphenyl (PCB No. 108) 35 µg/ml in isooctane
  • Formula:
    C₁₂H₅Cl₅
  • Description:
    2,3,3',4,5'-Pentachlorobiphenyl (PCB No. 108)