Organic reference standard, 2,2',5-Trichlorobiphenyl (PCB No. 18) 35 µg/ml in isooctane
Catalog # ACCUC-018S
Supplier: AccuStandard
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:37680-65-2
- Description:2,2',5-Trichlorobiphenyl (PCB No. 18) 35 µg/ml in isooctane
- Formula:C₁₂H₇Cl₃
- Description:2,2',5-Trichlorobiphenyl (PCB No. 18)