Order Entry
Northern Ireland
ContactUsLinkComponent
Potassium-D-gluconate for synthesis, Sigma-Aldrich®
Potassium-D-gluconate for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Potassium-D-gluconate for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    590 g/l (20 °C)
  • Melting Point:
    174 - 176 °C
  • Molar Mass:
    234.24 g/mol
  • Bulk Density:
    600 kg/m3
  • pH:
    7.0 - 8.3 (100 g/l, H2O, 20 °C)
  • Ignition Point:
    200 °C

Specification Test Results

Related Information

Frequently Bought Together