Organic reference standard, 2,2',4,6-Tetrachlorobiphenyl (PCB No. 50) 35 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-50
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:62796-65-0
- Description:2,2',4,6-Tetrachlorobiphenyl (PCB No. 50) 35 µg/ml in isooctane
- Formula:C₁₂H₆Cl₄
- Description:2,2',4,6-Tetrachlorobiphenyl (PCB No. 50)