Order Entry
Northern Ireland
ContactUsLinkComponent
 
L(+)-Lithium lactate
Catalog # MOLEM33846384
Supplier:  Molekula
CAS Number:  
L(+)-Lithium lactate
Catalog # MOLEM33846384
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :96.01 g/mol
Boiling Pt:200 °C (1013 hPa)
Melting Pt:75 °C
Storage Temperature:Ambient
EINECS:248-692-5
CAS Number:27848-80-2
Formula:C₃H₅LiO₃
MDL Number:MFCD00065512