Organic reference standard, 2,2',3,4,5',6-Hexachlorobiphenyl (PCB No. 144) 35 µg/ml in isooctane, CIL
PROOCIL-PCB-144
:
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:68194-14-9
- Description:2,2',3,4,5',6-Hexachlorobiphenyl (PCB No. 144) 35 µg/ml in isooctane
- Formula:C₁₂H₄Cl₆
- Description:2,2',3,4,5',6-Hexachlorobiphenyl (PCB No. 144)