ContactUsLinkComponent
VWR®, Filter Tips, Next Generation Refill, Low Retention
VWR®, Filter Tips, Next Generation Refill, Low Retention
Catalog # 732-3350
Supplier:  VWR Collection
undefined
VWR®, Filter Tips, Next Generation Refill, Low Retention
Catalog # 732-3350
Supplier:  VWR Collection
Specifications
  • CE-IVD:
    +
  • Packed:
    10 refills/pack (Next Generation)
  • Sterile:
    +
  • Volume:
    10 µl
  • Seq:
    02
  • Environmentally Preferable:
  • Tip style:
    Extended length, low retention, with TubeGuard
  • Pk:
    960
Specifications
About this item
These aerosol filter tips, with their patented filters made from high molecular weight polyethylene, offer complete protection from sample carry-over. Ideal for PCR and DNA manipulation.
  • Extremely low surface tension significantly reduces sample loss and provides substantially higher reproducibility when working with critical media
  • Easy to use refill system - new pipette tip racks, inserts and transfer cover preventing tip wobbling during refill and keeping tips clean
  • 95% renewable materials - compostable tip base from organic non petroleum resin, carton made from 100% recycled fibres, printing is soy-based vegetable ink
  • Certified human DNA-/RNase-/DNase-free
  • Free from pyrogens, ATP and protease activity
Next Generation is a complete system designed with a low carbon footprint that includes the first ever use of bio-plastic (non petroleum) compostable resin in pipette tip refills. Next Generation’s small footprint extends to the package, with a reclosable box that helps maximise storage space in crowded labs.