Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Northern Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(-)-Dimethyl D-tartrate
(-)-Dimethyl-D-tartrate for synthesis
(-)-Dimethyl-D-tartrate for synthesis
(-)-Dimethyl-D-tartrate for synthesis
8.18723.0010
:
Merck
:
(-)-Dimethyl-D-tartrate for synthesis
8.18723.0010
:
Merck
:
Pack type:
Glass bottle for solids
Pk:
10 g
Seq:
3236
:
:
178.14 g/mol
:
245 °C (1013 hPa)
:
45…48 °C
:
236-118-6
:
13171-64-7
:
C₆H₁₀O₆
:
MFCD00008445