Organic reference standard, 2,2',3,4,5,5',6-Heptachlorobiphenyl (PCB No. 185) 35 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-185
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:52712-05-7
- Description:2,2',3,4,5,5',6-Heptachlorobiphenyl (PCB No. 185) 35 µg/ml in isooctane
- Formula:C₁₂H₃Cl₇
- Description:2,2',3,4,5,5',6-Heptachlorobiphenyl (PCB No. 185)