Organic reference standard, o-Xylene 200 µg/ml in methanol
Catalog # 123352D
Supplier: AccuStandard
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:95-47-6
- Description:o-Xylene 200 µg/ml in methanol
- Formula:C₈H₁₀
- Description:o-Xylene