Organic reference standard, Trichlorofluoromethane 200 µg/ml in methanol
Catalog # 123512B
Supplier: AccuStandard
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:75-69-4
- Description:Trichlorofluoromethane 200 µg/ml in methanol
- Formula:CFCl₃
- Description:Trichlorofluoromethane