
Iron(II) sulphate heptahydrate ≥99.999% (metals basis), Puratronic®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7782-63-0
:
Formula:FeSO₄·7H₂O
MDL Number:MFCD00149719
:278.02 g/mol
Merck Index:14,4057
Melting Pt:64 °C (-6H₂O)
Density:1,897
EINECS:231-753-5
:GHS07
Storage Temperature:Ambient
Frequently Bought Together