




Nickel(II) nitrate hexahydrate ≥99.9985% (metals basis), Puratronic®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13478-00-7
:
Formula:Ni(NO₃)₂·6H₂O
MDL Number:MFCD00149805
:290.79 g/mol
Merck Index:14,6513
Boiling Pt:137 °C
Melting Pt:56,7 °C
Density:2,05
EINECS:236-068-5
UN:2725
ADR:5.1,III
:GHS03
:GHS08
:GHS05
:GHS07
:GHS09
Storage Temperature:Ambient
Frequently Bought Together