
Iridium(III) chloride hydrate ≥99.8% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:14996-61-3
:MFCD00149749
Formula:Cl₃Ir·xH₂O
MDL Number:MFCD00149750
Merck Index:14,5088
Density:5,3 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:233-044-6
UN:3260
ADR:8,II
:298.58 g/mol (anhy)
:GHS05
Related Information
Frequently Bought Together