

Yttrium (III) nitrate hydrate ≥99.99% (REO, rare earth oxide basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:140191-40-8
:13494-98-9
Formula:N₃O₉Y·H₂O
MDL Number:MFCD00011469
:274.92 g/mol (anhy)
Merck Index:14,10107
Storage Temperature:Ambient
EINECS:233-802-6
UN:1477
ADR:5.1,III
:GHS03
:GHS07
Frequently Bought Together